86879 Fluka

Tetrabutylammonium hexafluorophosphate

for electrochemical analysis, ≥99.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 3109-63-5

  • Linear Formula (CH3CH2CH2CH2)4N(PF6)

  • Molecular Weight 387.43

  •  Beilstein Registry Number 4056584

  •  EC Number 221-472-6

  •  MDL number MFCD00011748

  •  PubChem Substance ID 24888801

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
assay   ≥99.0% (CHN)
  ≥99.0%
mp   244-246 °C(lit.)
solubility   acetonitrile: soluble0.1 g/mL, clear, colorless

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Supporting electrolyte soluble in organic solvents which is very stable1

Price and Availability

Customers Also Viewed

Acetonitrile

anhydrous, 99.8%

Ferrocene

98%

Tetrabutylammonium hexafluorophosphate

purum, ≥98.0% (CHN)

Tetrabutylammonium perchlorate

for electrochemical analysis, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?