86880 Fluka

Tetrabutylammonium hydroxide solution

technical, ~40% in H2O (~1.5 M)

DOWNLOAD MSDS (PDF)
  • CAS Number 2052-49-5

  • Linear Formula (CH3CH2CH2CH2)4N(OH)

  • Molecular Weight 259.47

  •  Beilstein Registry Number 3571228

  •  MDL number MFCD00009425

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Organic Bases, Research Essentials,
grade   technical
concentration   ~40% in H2O (~1.5 M)
impurities   ≤3% halides (as bromide)
anion traces   sulfate (SO42-): ≤3000 mg/kg

Description

Caution

may crystallize; can be redissolved on heating to 40°C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Tetrabutylammonium hydroxide solution

~40% in H2O, for ion chromatography

Tetrabutylammonium hydroxide solution

purum, ~40% in H2O (~1.5 M)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?