• USA Home
  • 86894 - Tetrabutylammonium iodide

EMAIL THIS PAGE TO A FRIEND

86894 Fluka

Tetrabutylammonium iodide

purum, ≥98.0% (AT)

Synonym: TBAI

  • CAS Number 311-28-4

  • Linear Formula (CH3CH2CH2CH2)4N(I)

  • Molecular Weight 369.37

  •  Beilstein Registry Number 3916152

  •  EC Number 206-220-5

  •  MDL number MFCD00011636

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
grade   purum
assay   ≥98.0% (AT)
mp   141-143 °C(lit.)
  143-147 °C

Description

Other Notes

This product has been replaced by 140775-ALDRICH | Tetrabutylammonium iodide reagent grade, 98%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Tetrabutylammonium iodide

for ion pair chromatography, ≥99.0%

Tetrabutylammonium iodide

for electrochemical analysis, ≥99.0%

Tetrabutylammonium iodide

≥99.0% (AT)

Tetrabutylammonium iodide

reagent grade, 98%

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36
WGK Germany 
3
RTECS 
BS5450000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Efficient [WO4](2-)-catalyzed chemical fixation of carbon dioxide with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Kimura T, Sunaba H, Kamata K, et al. Inorg. Chem. 51(23), 13001-8, (2012)

Loading...


A highly negatively charged γ-Keggin germanodecatungstate efficient for Knoevenagel condensation. Sugahara K, Kimura T, Kamata K, et al. Chem. Commun. (Camb.) 48(67), 8422-4, (2012)

Loading...


Rhodium acetate/base-catalyzed N-silylation of indole derivatives with hydrosilanes. Itagaki S, Kamata K, Yamaguchi K, et al. Chem. Commun. (Camb.) 48(74), 9269-71, (2012)

Loading...


Ultrafast photo-induced charge transfer unveiled by two-dimensional electronic spectroscopy. Bixner O, Lukeš V, Mančal T, et al. J. Chem. Phys. 136(20), 204503, (2012)

Loading...


High-turnover hypoiodite catalysis for asymmetric synthesis of tocopherols. Uyanik M, Hayashi H, and Ishihara K Science 345(6194), 291-4, (2014)

Loading...


pH-dependent promotion of phospholipid flip-flop by the KcsA potassium channel. Nakao H, Ikeda K, Iwamoto M, et al. Biochim. Biophys. Acta 1848(1 Pt A), 145-50, (2015)

Loading...


Rapid and efficient synthesis of nucleoside polyphosphates and their conjugates using sulfonyl imidazolium salts. Mohamady S and Taylor SD Curr. Protoc. Nucleic Acid Chem. Chapter 13, Unit13.11, (2012)

Loading...


Convenient and efficient approach to the permanent or reversible conjugation of RNA and DNA sequences with functional groups. Cieślak J, Ausín C, Grajkowski A, et al. Curr. Protoc. Nucleic Acid Chem. Chapter 4, Unit4.52, (2012)

Loading...


Tetrabutylammonium iodide catalyzed synthesis of allylic ester with tert-butyl hydroperoxide as an oxidant. Shi E, Shao Y, Chen S, et al. Org. Lett. 14(13), 3384-7, (2012)

Loading...


The voltage-dependent gate in MthK potassium channels is located at the selectivity filter. Posson DJ, McCoy JG, and Nimigean CM Nat. Struct. Mol. Biol. 20(2), 159-66, (2013)

Loading...


Tandem cyclization of α-cyano α-alkynyl aryl ketones induced by tert-butyl hydroperoxide and tetrabutylammonium iodide. Wong YC, Tseng CT, Kao TT, et al. Org. Lett. 14(23), 6024-7, (2012)

Loading...


Tetrabutylammonium 2-pyridyltriolborate salts for Suzuki-Miyaura cross-coupling reactions with aryl chlorides. Sakashita S, Takizawa M, Sugai J, et al. Org. Lett. 15(17), 4308-11, (2013)

Loading...


Novel reverse-phase ion pair-high performance liquid chromatography separation of heparin, heparan sulfate and low molecular weight-heparins disaccharides and oligosaccharides. Galeotti F and Volpi N J. Chromatogr. A 1284, 141-7, (2013)

Loading...


IBS-catalyzed regioselective oxidation of phenols to 1,2-quinones with Oxone®. Uyanik M, Mutsuga T, and Ishihara K Molecules 17(7), 8604-16, (2012)

Loading...


Synthesis of 2-nitroglycals from glycals using the tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine reagent system and base-catalyzed Ferrier rearrangement of acetylated 2-nitroglycals. Dharuman S, Gupta P, Kancharla PK, et al. J. Org. Chem. 78(17), 8442-50, (2013)

Loading...


Quantitative imaging of ion transport through single nanopores by high-resolution scanning electrochemical microscopy. Shen M, Ishimatsu R, Kim J, et al. J. Am. Chem. Soc. 134(24), 9856-9, (2012)

Loading...


Photoinduced ligand exchange and DNA binding of cis-[Ru(phpy)(phen)(CH3CN)2]+ with long wavelength visible light. Sears RB, Joyce LE, Ojaimi M, et al. J. Inorg. Biochem. 121, 77-87, (2013)

Loading...


Thermal-induced dynamic self-assembly of adenine-grafted polyoxometalate complexes. He Z, Yan Y, Li B, et al. Dalton Trans. 41(33), 10043-51, (2012)

Loading...


Development of a new and environment friendly hollow fiber-supported liquid phase microextraction using vesicular aggregate-based supramolecular solvent. Moradi M, Yamini Y, Rezaei F, et al. Analyst 137(15), 3549-57, (2012)

Loading...


Wheat straw cellulose dissolution and isolation by tetra-n-butylammonium hydroxide. Zhong C, Wang C, Huang F, et al. Carbohydr. Polym. 94(1), 38-45, (2013)

Loading...


Monitoring of thiopurine metabolites - a high-performance liquid chromatography method for clinical use. Vikingsson S, Almer S, Peterson C, et al. J. Pharm. Biomed. Anal. 75, 145-52, (2013)

Loading...


Extraction and stability of pesticide multiresidues from natural water on a mixed-mode admicellar sorbent. Luque N and Rubio S J. Chromatogr. A 1248, 74-83, (2012)

Loading...


Gelation via ion exchange in discotic suspensions. Chang YW, Mejia AF, Cheng Z, et al. Phys. Rev. Lett. 108(24), 247802, (2012)

Loading...


Synthetic chloride transporters with the binding mode observed in a ClC chloride channel. Choi YR, Chae MK, Kim D, et al. Chem. Commun. (Camb.) 48(83), 10346-8, (2012)

Loading...


Molecular and system parameters governing mass and charge transport in polar liquids and electrolytes. Petrowsky M, Fleshman A, Ismail M, et al. J. Phys. Chem. B 116(33), 10098-105, (2012)

Loading...


A series of amino acid functionalized tripodal hexaamide anion receptors: ion-pair-assisted capped-cleft formation by a pentafluorophenyl-functionalized amide. Bose P, Ravikumar I, Akhuli B, et al. Chem. Asian J. 7(10), 2373-80, (2012)

Loading...


Small-angle neutron scattering study of temperature vs. salt dependence of clouding in charged micellar system. Chinchalikar AJ, Aswal VK, Kohlbrecher J, et al. Eur. Phys. J. E 35(7), 55, (2012)

Loading...


Development of a novel expression, ZI MAX/K ZI, for determination of the counter-anion effect on the antimicrobial activity of tetrabutylammonium salts. Michelle L Ingalsbe et al Bioorg. Med. Chem. Lett. 19, 4984-7, (2009)

Loading...


Adsorption of phenanthrene by quaternary ammonium surfactant modified peat and the mechanism involved. Zhou YB, Chen L, Wang XQ, et al. Water Sci. Technol. 66(4), 810-5, (2012)

Loading...


RNA editing in eag potassium channels: biophysical consequences of editing a conserved S6 residue. Ryan MY, Maloney R, Fineberg JD, et al. Channels (Austin.) 6(6), 443-52, (2012)

Loading...


Synthesis of 9,9'-[1,2-ethanediylbis(oxymethylene)]bis-2-amino-1,9-dihydro-6H-purin-6-one, an impurity of acyclovir. Suárez RM, Matía MP, Novella JL, et al. Molecules 17(8), 8735-41, (2012)

Loading...


Beil. 4,IV,558

Fieser 5,646 / Fieser 6,566 / Fieser 7,355 / Fieser 9,447 / Fieser 11,501 / Fieser 13,287

Aldrich MSDS 1, 1645:D / Corp MSDS 1 (2), 3238:C / FT-IR 1 (1), 391:A / FT-IR 2 (1), 611:D / IR-Spectra (2), 205:F / IR-Spectra (3), 230:B / NMR-Reference 2 (1), 345:B / RegBook 1 (1), 413:G / Sax 6, 2514 / Sigma FT-IR 1 (2), 904:C / Structure Index 1, 60:D:3

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?