• USA Home
  • 86896 - Tetrabutylammonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND

86896 Fluka

Tetrabutylammonium tetrafluoroborate

for electrochemical analysis, ≥99.0%

Purchase

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
assay   ≥99.0% (T)
  ≥99.0%
mp   155-161 °C(lit.)
  159-162 °C
solubility   methanol: soluble0.1 g/mL, clear, colorless

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Tetrabutylammonium hexafluorophosphate

for electrochemical analysis, ≥99.0%

Tetrabutylammonium perchlorate

for electrochemical analysis, ≥99.0%

Tetrabutylammonium tetrafluoroborate

puriss., ≥99.0% (T)

Tetraethylammonium tetrafluoroborate

for electrochemical analysis, ≥99.0%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-39
WGK Germany 
3
Flash Point(F) 
>199.4 °F
Flash Point(C) 
>93 °C

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?