Synonym: n-Tetracontane
CAS Number 4181-95-7
Linear Formula CH3(CH2)38CH3
Molecular Weight 563.08
Beilstein Registry Number 1800152
EC Number 224-055-7
MDL number MFCD00015267
PubChem Substance ID 24888850
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Hydrocarbons, |
| grade | analytical standard |
| assay | ≥98.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 80-83 °C |
| 80-84 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?