Synonym:
γ-Nonyl-γ-valerolactone,
6-
CAS Number 92446-06-5
Empirical Formula (Hill Notation) C14H26O2
Molecular Weight 226.36
Beilstein Registry Number 1619271
MDL number MFCD00059809
PubChem Substance ID 24888722
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Heterocyclics, |
| assay | ≥96.0% (GC) |
| density | 0.936 g/mL at 20 °C(lit.) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?