• USA Home
  • 87208 - Myristyltrimethylammonium bromide

EMAIL THIS PAGE TO A FRIEND

87208 Fluka

Myristyltrimethylammonium bromide

for ion pair chromatography, ≥99.0% (AT)

Synonym: Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide

Purchase

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Buffers & Reagents,
grade   for ion pair chromatography
description   cationic
assay   ≥99.0% (AT)
mol wt   micellar avg mol wt 27,000
aggregation number   80
CMC   4-5 mM(20-25°C)
mp   245-250 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 240 nm Amax: ≤0.08
  λ: 250 nm Amax: ≤0.05
  λ: 260 nm Amax: ≤0.04
  λ: 500 nm Amax: ≤0.02
suitability   corresponds to standard for filter test

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Customers Also Viewed

Decamethonium bromide

for ion pair chromatography, ≥99.0% (AT)

Dodecyltrimethylammonium bromide

for ion pair chromatography, ≥98.5% (AT)

Trimethyltetradecylammonium bromide

European Pharmacopoeia (EP) Reference Standard

Trimethyltetradecylammonium hydrogen sulfate

for ion pair chromatography, ≥99.0% (T)

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
C
Risk Statements (Europe) 
34
Safety Statements (Europe) 
26-36/37/39-45
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3
RTECS 
BS5776000

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Small molecule inhibitors of aggregation indicate that amyloid beta oligomerization and fibrillization pathways are independent and distinct. Mihaela Necula et al J. Biol. Chem. 282, 10311-24, (2007)

Loading...


Spectroscopic study of tetradecyltrimethylammonium bromide Pt-C14TAB nanoparticles: structure and stability. Borodko Y, Jones L, Lee H, et al. Langmuir 25(12), 6665-71, (2009)

Loading...


The dynamin inhibitors MiTMAB and OcTMAB induce cytokinesis failure and inhibit cell proliferation in human cancer cells. Joshi S, Perera S, Gilbert J, et al. Mol. Cancer Ther. 9(7), 1995-2006, (2010)

Loading...


Flotation of marine microalgae: effect of algal hydrophobicity. Garg S, Li Y, Wang L, et al. Bioresour. Technol. 121, 471-4, (2012)

Loading...


Characterisation of organoclays and adsorption of p-nitrophenol: environmental application. Park Y, Ayoko GA, and Frost RL J. Colloid. Interface Sci. 360(2), 440-56, (2011)

Loading...


In-channel amperometric detection for microchip electrophoresis using a wireless isolated potentiostat. Gunasekara DB, Hulvey MK, and Lunte SM Electrophoresis 32(8), 832-7, (2011)

Loading...


Foam invasion through a single pore. Delbos A and Pitois O Phys. Rev. E. Stat. Nonlin. Soft Matter Phys. 84(1 Pt 1), 011404, (2011)

Loading...


Comprehensive study of tartrazine/cationic surfactant interaction. Shahir AA, Javadian S, Razavizadeh BB, Gharibi H. J. Phys. Chem. B 115, 14435-14444, (2011)

Loading...


Selectivity patterns in micellar electrokinetic chromatography: characterization of fluorinated and aliphatic alcohol modifiers by micellar selectivity triangle. Fu C and Khaledi MG J. Chromatogr. A 1216(10), 1901-7, (2009)

Loading...


Phase behaviors and self-assembly properties of two catanionic surfactant systems: C(8)F(17)COOH/TTAOH/H(2)O and C(8)H(17)COOH/TTAOH/H(2)O. Zhang J, Song A, Li Z, et al. J. Phys. Chem. B 114(41), 13128-35, (2010)

Loading...


Phase behavior and rheological properties of salt-free catanionic surfactant mixtures in the presence of bile acids. Liu C, Hao J, and Wu Z J. Phys. Chem. B 114(30), 9795-804, (2010)

Loading...


Regulation of the retention factor for weak acids in micellar electrokinetic chromatography with cationic surfactant via variation of the chloride concentration. Orentaitė I, Maruška A, and Pyell U Electrophoresis 32(5), 604-13, (2011)

Loading...


Macromolecular and morphological evolution of poly(styrene sulfonate) complexes with tetradecyltrimethylammonium bromide. Popov A, Zakharova J, Wasserman A, et al. J. Phys. Chem. B 116(40), 12332-40, (2012)

Loading...


New tetradecyltrimethylammonium-selective electrodes: surface composition and topography as correlated with electrode's life span. Marafie HM, Al-Shammari TF, and Shoukry AF Talanta 91, 26-32, (2012)

Loading...


An investigation of drug binding ability of a surface active ionic liquid: micellization, electrochemical, and spectroscopic studies. Mahajan S, Sharma R, and Mahajan RK Langmuir 28(50), 17238-46, (2012)

Loading...


Spectroscopic studies on the interaction of cationic surfactants with bovine serum albumin. Gull N, Chodankar S, Aswal VK, et al. Colloids Surf. B Biointerfaces 69(1), 122-8, (2009)

Loading...


NMR study of the dynamics of cationic gemini surfactant 14-2-14 in mixed solutions with conventional surfactants. Jiang Y, Lu XY, Chen H, et al. J. Phys. Chem. B 113(24), 8357-61, (2009)

Loading...


Structural studies of surfactants at the oil-water interface by neutron reflectometery. Zarbakhsh A, Webster JR, and Eames J Langmuir 25(7), 3953-6, (2009)

Loading...


Unfolding of rabbit serum albumin by cationic surfactants: surface tensiometry, small-angle neutron scattering, intrinsic fluorescence, resonance Rayleigh scattering and circular dichroism studies. Ali MS, Gull N, Khan JM, et al. J. Colloid. Interface Sci. 352(2), 436-43, (2010)

Loading...


Spectroscopic studies on the competitive interaction between polystyrene sodium sulfonate with polycations and the N-tetradecyl trimethyl ammonium bromide surfactant. Estrela-Lopis I, Iturri Ramos JJ, Donath E, et al. J. Phys. Chem. B 114(1), 84-91, (2010)

Loading...


Phase stability of Keplerate-type polyoxomolybdates controlled by added cationic surfactant. Fan D and Hao J J. Colloid. Interface Sci. 333(2), 757-63, (2009)

Loading...


Pseudomonas putida A ATCC 12633 oxidizes trimethylamine aerobically via two different pathways. Liffourrena AS, Salvano MA, and Lucchesi GI Arch. Microbiol. 192(6), 471-6, (2010)

Loading...


Photoreversible polymer-surfactant micelles using the molecular recognition of α-cyclodextrin. Dong ZQ, Cao Y, Han XJ, et al. Langmuir 29(10), 3188-94, (2013)

Loading...


Solubilization of n-alkylbenzene and n-perfluoroalkylbenzene in hydrogenated and fluorinated surfactants micelles. Matsuoka K, Takeuchi E, Takahashi M, et al. J. Colloid. Interface Sci. 333(2), 641-5, (2009)

Loading...


Effect of ionic surfactants on ion vibration current and colloid vibration current. Takata Y, Shimatsu Y, Nagahashi T, et al. J. Oleo Sci. 59(8), 401-6, (2010)

Loading...


Physiological role of phosphatidylcholine in the Pseudomonas putida A ATCC 12633 response to tetradecyltrimethylammonium bromide and aluminium. Boeris PS, Liffourrena AS, Salvano MA, et al. Lett. Appl. Microbiol. 49(4), 491-6, (2009)

Loading...


Merck 14,6336

Beil. 4,IV,813

Aldrich MSDS 1, 1332:C / Corp MSDS 1 (2), 2483:B / FT-IR 1 (1), 392:C / FT-IR 2 (1), 618:A / IR-Spectra (3), 230:D / RegBook 1 (1), 419:G / Sigma FT-IR 1 (2), 67:D / Structure Index 1, 61:C:4

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?