EMAIL THIS PAGE TO A FRIEND

87367 Fluka

Tetrahydrofuran

for HPLC, ≥99.8% (GC)

  • CAS Number 109-99-9

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 102391

  •  EC Number 203-726-8

  •  MDL number MFCD00005356

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   for HPLC
vapor density   2.5 (vs air)
vapor pressure   114 mmHg ( 15 °C)
  143 mmHg ( 20 °C)
assay   ≥99.8% (GC)
autoignition temp.   610 °F
shelf life   limited shelf life, expiry date on the label
expl. lim.   11.8 %
impurities   ≤0.02% water
evapn. residue   ≤0.001%
refractive index   n20/D 1.407(lit.)
  n20/D 1.407
bp   65-67 °C(lit.)
mp   −108 °C(lit.)
density   0.889 g/mL at 25 °C(lit.)
solvent strength ε (Al2O3)   0.45
λ   1 cm path, H2O reference
UV absorption   λ: 230 nm Amax: 1.0
  λ: 270 nm Amax: 0.1
  λ: 310 nm Amax: 0.01

Price and Availability

Customers Also Viewed

Tetrahydrofuran

inhibitor-free, CHROMASOLV® Plus, for HPLC, ≥99.9%

Tetrahydrofuran

CHROMASOLV® Plus, for HPLC, ≥99.9%, inhibitor-free

Tetrahydrofuran

analytical standard

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
13-16-29-33-36-37-46
RIDADR 
UN 2056 3 / PGII
WGK Germany 
1
RTECS 
LU5950000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Protocols & Articles

Related Products

recommended

Product #

Description

Add to Cart

Z683299 XPell indicating pellets for peroxides prevention, CE for organic solvents

replaced by

Product #

Description

Add to Cart

34865 Tetrahydrofuran, inhibitor-free, CHROMASOLV® Plus, for HPLC, ≥99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?