87639 Fluka

5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate)

for fluorescence, ≥95% (N)

DOWNLOAD MSDS (PDF)

Synonym: meso-Tetra(N-methyl-4-pyridyl)porphine tetratosylate salt, TMPyP

  • CAS Number 36951-72-1

  • Empirical Formula (Hill Notation) C72H66N8O12S4

  • Molecular Weight 1363.60

  •  Beilstein Registry Number 6049195

  •  MDL number MFCD00013468

  •  PubChem Substance ID 24888923

Properties

grade   for fluorescence
assay   ≥95% (N)
fluorescence   λex 520 nm; λem ~700 nm in 0.1 M Tris pH 8.0
storage temp.   −20°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?