87708 Fluka

Tetramethylammonium bromide

for ion pair chromatography, ≥99.0% (AT)

DOWNLOAD MSDS (PDF)
  • CAS Number 64-20-0

  • Linear Formula (CH3)4N(Br)

  • Molecular Weight 154.05

  •  Beilstein Registry Number 3620955

  •  EC Number 200-581-2

  •  MDL number MFCD00011626

  •  PubChem Substance ID 24888934

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (AT)
mp   >300 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 240 nm Amax: 0.06
  λ: 250 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Tetramethylammonium bromide

ACS reagent, ≥98.0%

Tetramethylammonium bromide

for electrochemical analysis, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?