87709 Fluka

Tetramethylammonium bromide

for electrochemical analysis, ≥99.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 64-20-0

  • Linear Formula (CH3)4N(Br)

  • Molecular Weight 154.05

  •  Beilstein Registry Number 3620955

  •  EC Number 200-581-2

  •  MDL number MFCD00011626

  •  PubChem Substance ID 24888935

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
assay   ≥99.0% (AT)
  ≥99.0%
mp   >300 °C(lit.)
solubility   H2O: soluble0.1 g/mL, clear, colorless

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Tetraethylammonium bromide

for electrochemical analysis, ≥99.0%

Tetramethylammonium bromide

for ion pair chromatography, ≥99.0% (AT)

Tetramethylammonium bromide

ACS reagent, ≥98.0%

Tetramethylammonium chloride solution

for molecular biology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?