87719 Fluka

Tetramethylammonium chloride

for electrochemical analysis, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: TMA

  • CAS Number 75-57-0

  • Linear Formula (CH3)4N(Cl)

  • Molecular Weight 109.60

  •  Beilstein Registry Number 3911201

  •  EC Number 200-880-8

  •  MDL number MFCD00011628

  •  PubChem Substance ID 24888937

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
assay   ≥99.0% (AT)
  ≥99.0%
mp   >300 °C(lit.)
solubility   methanol: soluble0.1 g/mL, clear, colorless

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Customers Also Viewed

Tetraethylammonium chloride

for electrochemical analysis, ≥99.0%

Tetramethylammonium chloride

BioUltra, for molecular biology, ≥99.0% (AT)

Tetramethylammonium chloride

for ion pair chromatography, ≥99.0% (AT)

Tetramethylammonium chloride

purum, ≥98.0% (AT)

Tetramethylammonium chloride

reagent grade, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?