87724 Fluka

Tetramethylammonium bisulfate

for ion pair chromatography, ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Tetramethylammonium hydrogen sulfate

  • CAS Number 80526-82-5

  • Linear Formula (CH3)4N(HSO4)

  • Molecular Weight 171.22

  •  Beilstein Registry Number 4829202

  •  EC Number 279-490-5

  •  MDL number MFCD00036149

  •  PubChem Substance ID 24888939

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (T)
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: 0.05
  λ: 220 nm Amax: 0.04
  λ: 230 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Price and Availability

Customers Also Viewed

Tetrabutylammonium bisulfate

for ion pair chromatography, ≥99.0%

Tetramethylammonium sulfate

for ion pair chromatography, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?