87729 Fluka

Tetramethylammonium hydroxide solution

purum, ~25% in H2O

DOWNLOAD MSDS (PDF)
  • CAS Number 75-59-2

  • Linear Formula (CH3)4N(OH)

  • Molecular Weight 91.15

  •  Beilstein Registry Number 3558708

  •  MDL number MFCD00008280

  • Popular Documents:  RAMAN FTIR (PDF)  

Properties

Related Categories Chemical Synthesis, Organic Bases, Synthetic Reagents More...
grade   purum
concentration   ~25% in H2O
impurities   ≤0.1% halides (as chloride)
refractive index   n20/D 1.382
anion traces   sulfate (SO42-): ≤1000 mg/kg

Price and Availability

Customers Also Viewed

Tetramethylammonium hydroxide solution

25 wt. % in methanol

Tetramethylammonium hydroxide solution

25% in H2O, TraceSELECT® Ultra

Tetramethylammonium hydroxide solution

25% in H2O, TraceSELECT®, for trace analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?