• USA Home
  • 87797 - 2,4,6,8-Tetramethylcyclotetrasiloxane

EMAIL THIS PAGE TO A FRIEND

87797 Fluka

2,4,6,8-Tetramethylcyclotetrasiloxane

technical, ≥95% (GC)

Synonym: TMCTS

Purchase

Properties

grade   technical
assay   ≥95% (GC)
form   liquid
refractive index   n20/D 1.387(lit.)
bp   134 °C(lit.)
mp   −69 °C(lit.)
density   0.986 g/mL at 25 °C(lit.)

Description

Features and Benefits

Precursor for gate dielectrics in thin-film transistors (TFT),1 and is a component of photochemically formed SiOX monolayers on TiO.2

Price and Availability

Customers Also Viewed

2,4,6,8-<WBR>Tetramethylcyclotetrasiloxane

≥99.5%, ≥99.999% trace metals basis

2,4,6,8-<WBR>Tetramethylcyclotetrasiloxane

packaged for use in deposition systems

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Risk Statements (Europe) 
10
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
75.2 °F
Flash Point(C) 
24 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. Wang,A. W. et al. Mater. Res. Soc. Symp. Proc. 424, 281, (1997)

2. Tada, H.; Tanaka, M. Thin Solid Films 281, 404, (1996)

Beil. 4,IV,4099

FT-IR 2 (3), 4387:D / Structure Index 1, 471:C:5

Related Products

replaced by

Product #

Description

Add to Cart

512990 2,4,6,8-Tetramethylcyclotetrasiloxane, ≥99.5%, ≥99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?