EMAIL THIS PAGE TO A FRIEND

87850 Fluka

Tetramethylurea

purum, ≥99.0% (GC)

Synonym: 1,1,3,3-Tetramethylurea, TMU

  • CAS Number 632-22-4

  • Linear Formula (CH3)2NCON(CH3)2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 773898

  •  EC Number 211-173-9

  •  MDL number MFCD00008319

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.451(lit.)
  n20/D 1.451
bp   177 °C(lit.)
  62-63 °C/12 mmHg(lit.)
mp   −1 °C(lit.)
density   0.968 g/mL at 20 °C(lit.)

Description

Other Notes

This product has been replaced by T24503-ALDRICH | Tetramethylurea 99%

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
YU2625000
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,9230

Beil. 4,IV,225

Fieser 1,1146 / Fieser 15,305

Aldrich MSDS 1, 1687:C / Corp MSDS 1 (2), 3305:A / FT-IR 1 (1), 804:A / FT-IR 2 (1), 1364:B / IR-Spectra (3), 476:F / IR-Spectra (2), 422:F / NMR-Reference 2 (1), 673:D / RegBook 1 (1), 943:C / Sax 6, 2549 / Sigma FT-IR 1 (2), 918:A / Structure Index 1, 146:A:8 / Vapor Phase 3, 790:D

Related Products

replaced by

Product #

Description

Add to Cart

T24503 Tetramethylurea, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?