• USA Home
  • 87890 - N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND

87890 Fluka

N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride

≥97.0% (AT)

Synonym: TMPPD, Wurster’s reagent

Purchase

Description

Application

Test reagent in microbiology for the classification of cytochrome oxidase positive aerobic microorganisms.; For oxidase reagent, use a 1% aqueous solution of N,N,N′,N′-Tetramethyl-p-phenylenediamine dihydrochloride

Packaging

5, 25 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 13,IV,107

Aldrich MSDS 1, 1684:A / Corp MSDS 1 (2), 3298:D / FT-IR 1 (1), 1243:C / FT-IR 2 (2), 2106:C / FT-NMR 1 (2), 544:B / NMR-Reference 2 (1), 1051:D / RegBook 1 (1), 1435:D / Sax 6, 2547 / Sigma FT-IR 1 (2), 916:D / Structure Index 1, 224:C:2

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?