CAS Number 5856-66-6
Linear Formula CH3(CH2)52CH3
Molecular Weight 759.45
Beilstein Registry Number 9467386
MDL number MFCD00216751
PubChem Substance ID 24889027
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Hydrocarbons, |
| grade | analytical standard |
| assay | ≥98.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 94-97 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?