CAS Number 4499-86-9
Linear Formula (CH3CH2CH2)4N(OH)
Molecular Weight 203.36
Beilstein Registry Number 4207983
EC Number 224-800-6
MDL number MFCD00009360
| Related Categories | Chemical Synthesis, Organic Bases, Synthetic Reagents More... |
| grade | technical |
| concentration | ~20% in H2O (~1.0 M) |
| impurities | ≤3% halides (as bromide) |
| density | 1.00 g/mL at 20 °C |
| anion traces | sulfate (SO42-): ≤1000 mg/kg |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?