CAS Number 288-47-1
Empirical Formula (Hill Notation) C3H3NS
Molecular Weight 85.13
Beilstein Registry Number 103852
EC Number 206-021-3
MDL number MFCD00005315
PubChem Substance ID 24889087
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, C3 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More... |
| grade | purum |
| assay | ≥98.0% (GC) |
| refractive index |
n |
|
n | |
| bp | 117-118 °C(lit.) |
| density | 1.2 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?