EMAIL THIS PAGE TO A FRIEND

88491 Fluka

Thiobenzamide

purum, ≥98.0% (N)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
grade   purum
assay   ≥98.0% (N)
color   yellow
mp   113-117 °C(lit.)
  113-117 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
T
Risk Statements (Europe) 
25
Safety Statements (Europe) 
45
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
CV5860000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Hydrogen sulfide enhances ulcer healing in rats. Wallace, J.L., et al. FASEB J. 21, 4070-4076, (2007)

Loading...


Facile synthesis of nitriles via manganese oxide promoted oxidative dehydrosulfurization of primary thioamides. Yamaguchi K, Yajima K, and Mizuno N Chem. Commun. (Camb.) 48(91), 11247-9, (2012)

Loading...


2,2'-Dithiobisbenzamides and 2-benzisothiazolones, two new classes of antiretroviral agents: SAR and mechanistic considerations. Domagala JM, Gogliotti R, Sanchez JP, et al. Drug Des. Discov. 15(1), 49-61, (1997)

Loading...


Chemical Research in Toxicology. In this issue. Chem. Res. Toxicol. 21(7), 1320-1, (2008)

Loading...


Gender differences in hepatic expression of flavin-containing monooxygenase isoforms (FMO1, FMO3, and FMO5) in mice. Falls JG, Blake BL, Cao Y, et al. J. Biochem. Toxicol. 10(3), 171-7, (1995)

Loading...


Protein targets of reactive metabolites of thiobenzamide in rat liver in vivo. Ikehata K, Duzhak TG, Galeva NA, et al. Chem. Res. Toxicol. 21(7), 1432-42, (2008)

Loading...


Possible role of the acetone-inducible cytochrome P-450IIE1 in the metabolism and hepatotoxicity of thiobenzamide. Chieli E, Saviozzi M, Puccini P, et al. Arch. Toxicol. 64(2), 122-7, (1990)

Loading...


Potent inhibition of alcohol self-administration in alcohol-preferring rats by a κ-opioid receptor antagonist. Cashman JR and Azar MR J. Pharmacol. Exp. Ther. 350(1), 171-80, (2014)

Loading...


Role of lipoxygenase in xenobiotic metabolism: sulfoxidation of thiobenzamide by purified soybean lipoxygenase. Naidu AK and Kulkarni AP Res. Commun. Chem. Pathol. Pharmacol. 71(2), 175-88, (1991)

Loading...


Involvement of CYP3A1, 2B1, and 2E1 in C-8 hydroxylation and CYP 1A2 and flavin-containing monooxygenase in N-demethylation of caffeine; identified by using inducer treated rat liver microsomes that are characterized with testosterone metabolic patterns. Chung WG, Roh HK, Kim HM, et al. Chem. Biol. Interact. 113(1), 1-14, (1998)

Loading...


Covalent modification of microsomal lipids by thiobenzamide metabolites in vivo. Ji T, Ikehata K, Koen YM, et al. Chem. Res. Toxicol. 20(4), 701-8, (2007)

Loading...


Presence of flavin-containing monooxygenase in rat brain. Bhamre S and Ravindranath V Biochem. Pharmacol. 42(2), 442-4, (1991)

Loading...


Persistent chemopreventive effect of S-adenosyl-L-methionine on the development of liver putative preneoplastic lesions induced by thiobenzamide in diethylnitrosamine-initiated rats. Simile MM, Saviozzi M, De Miglio MR, et al. Carcinogenesis 17(7), 1533-7, (1996)

Loading...


Effects of the herbicide chlorthiamid on the olfactory mucosa. Eriksson C and Brittebo EB Toxicol. Lett. 76(3), 203-8, (1995)

Loading...


Placental peroxidase--further purification of the enzyme and oxidation of thiobenzamide. Joseph P, Srinivasan NS, and Kulkarni AP Placenta 14(3), 309-19, (1993)

Loading...


Structure-activity relationships for a series of thiobenzamide influenza fusion inhibitors derived from 1,3,3-trimethyl-5-hydroxy-cyclohexylmethylamine. Yu KL, Torri AF, Luo G, et al. Bioorg. Med. Chem. Lett. 12(23), 3379-82, (2002)

Loading...


Peroxidase-catalyzed S-oxygenation: mechanism of oxygen transfer for lactoperoxidase. Doerge DR, Cooray NM, and Brewster ME Biochemistry 30(37), 8960-4, (1991)

Loading...


Metabolism of thioamides by Ralstonia pickettii TA. Dodge AG, Richman JE, Johnson G, et al. Appl. Environ. Microbiol. 72(12), 7468-76, (2006)

Loading...


Relative hepatotoxicity of 2-(substituted phenyl)thiazoles and substituted thiobenzamides in mice: evidence for the involvement of thiobenzamides as ring cleavage metabolites in the hepatotoxicity of 2-phenylthiazoles. Mizutani T and Suzuki K Toxicol. Lett. 85(2), 101-5, (1996)

Loading...


Determination of FAD-binding domain in flavin-containing monooxygenase 1 (FMO1). Kubo A, Itoh S, Itoh K, et al. Arch. Biochem. Biophys. 345(2), 271-7, (1997)

Loading...


Flavin-containing monooxygenase activity can be inhibited by nitric oxide-mediated S-nitrosylation. Ryu SD, Yi HG, Cha YN, et al. Life Sci. 75(21), 2559-72, (2004)

Loading...


Thiobenzamide-induced hepatotoxicity: effects of substituents and route of administration on the nature and extent of liver injury. Grossman SJ, Patrick DH, Kornbrust D, et al. Toxicol. Appl. Pharmacol. 111(3), 388-408, (1991)

Loading...


Determination of p-methylthiobenzamide and p-methylthiobenzamide-S-oxide from rat plasma using solid-phase extraction and high-performance liquid chromatography. Davidson VP and Traiger GJ J. Chromatogr. A 567(1), 213-20, (1991)

Loading...


Induction of flavin-containing monooxygenase (FMO1) by a polycyclic aromatic hydrocarbon, 3-methylcholanthrene, in rat liver. Chung WG, Park CS, Roh HK, et al. Mol. Cells 7(6), 738-41, (1997)

Loading...


ETM-ANN approach application for thiobenzamide and quinolizidine derivatives. Saracoglu M, Kandemirli F, Kovalishyn V, et al. J. Biomed. Biotechnol. 2010, doi:10.1155/2010/693031, (2010)

Loading...


Conformational polymorphism in N-(4'-methoxyphenyl)- 3-bromothiobenzamide. Bashkirava A, Andrews PC, Junk PC, et al. Chem. Asian J. 2(4), 530-8, (2007)

Loading...


Beil. 9,IV,1376

Aldrich MSDS 1, 1697:D / Corp MSDS 1 (2), 3325:C / FT-IR 1 (2), 415:C / FT-IR 2 (2), 3094:A / FT-NMR 1 (2), 1472:C / IR-Spectra (3), 1106:D / IR-Spectra (2), 965:A / NMR-Reference 2 (2), 383:C / RegBook 1 (2), 2093:B / Structure Index 1, 331:B:2

Related Products

replaced by

Product #

Description

Add to Cart

148229 Thiobenzamide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?