88588 Fluka

Plate-Count-Agar according to Buchbinder et al.

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Casein-peptone Dextrose Yeast Agar according to Buchbinder et al., Standard Methods Agar according to Buchbinder et al., Tryptone Glucose Yeast Extract Agar according to Buchbinder et al.

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Bacillus, Bacteria (General Media),
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15 g/L
  casein enzymic hydrolysate, 5 g/L
  dextrose, 1 g/L
  yeast extract, 2.5 g/L
final pH   7.0±0.2 (25 °C)
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Enterococcus
  nonselective for Escherichia coli and Coliforms
  nonselective for Lactococci
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Plate-Count-Agar is recommended for the plate count of microrganisms in food, dairy products, water and waste water.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology, ready to use media in the bottle

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?