89050 Fluka

Thiosemicarbazide

puriss. p.a., ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Thiocarbamoyl hydrazide

  • CAS Number 79-19-6

  • Linear Formula NH2CSNHNH2

  • Molecular Weight 91.14

  •  Beilstein Registry Number 506320

  •  EC Number 201-184-7

  •  MDL number MFCD00007620

  •  PubChem Substance ID 24889170

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., T-Z, Puriss p.a. More...
grade   puriss. p.a.
assay   ≥99.0% (RT)
  ≥99.0%
ign. residue   ≤0.05% (as SO4)
mp   180-183 °C (dec.)(lit.)
anion traces   sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Customers Also Viewed

2,3-Butanedione monoxime

for spectrophotometric det. of urea, ≥99.0%

Thiosemicarbazide

purum, ≥98.0% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?