• USA Home
  • 89053 - Thiostrepton from Streptomyces azureus

EMAIL THIS PAGE TO A FRIEND

89053 Fluka

Thiostrepton from Streptomyces azureus

≥90% (HPLC)

Purchase

Properties

assay   ≥90% (HPLC)
impurities   ≤5% water
storage temp.   −20°C

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
22
WGK Germany 
3
RTECS 
XN6300100

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,9364

Related Products

replaced by

Product #

Description

Add to Cart

T8902 Thiostrepton from Streptomyces azureus, ≥90% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?