• USA Home
  • 89320 - 1-Hexyl-3-methylimidazolium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND

89320 Fluka

1-Hexyl-3-methylimidazolium hexafluorophosphate

≥97.0% (HPLC)

Synonym: HMIMPF6

  • CAS Number 304680-35-1

  • Empirical Formula (Hill Notation) C10H19F6N2P

  • Molecular Weight 312.24

  •  Beilstein Registry Number 8955409

  •  MDL number MFCD03093296

  •  PubChem Substance ID 24889211

Purchase

Properties

Related Categories Chemical Synthesis, Imidazolium, Ionic Liquids, Specialty Synthesis
assay   ≥97.0% (HPLC)
electrochemical window   5.5 V
refractive index   n20/D 1.419

Description

Other Notes

Ionic liquid employed in Friedel-Crafts alkylation of aromatic compounds;1 polarity of salt.2

Packaging

5 g in poly tube

50 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. C.E. Song et al. Chem. Commun. (Camb.), 1695, (2000)

2. A.J. Carmichael, K.R. Seddon J. Phys. Org. Chem. 13, 591, (2000)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?