CAS Number 107438-79-9
Empirical Formula (Hill Notation) C20H24O10
Molecular Weight 424.40
| Related Categories | Alphabetical, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| assay | ≥90% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?