• USA Home
  • 89730 - p-Toluenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND

89730 Fluka

p-Toluenesulfonyl chloride

ReagentPlus®, ≥99.0%

Synonym: Tosyl chloride

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents HPLC,
grade   ReagentPlus®
vapor pressure   1 mmHg ( 88 °C)
assay   ≥99.0%
bp   134 °C/10 mmHg(lit.)
mp   65-69 °C(lit.)
  66-69 °C
solubility   methylene chloride: soluble0.2 g/mL, clear

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Explore the New Analytical Standards Portal

Additional Products to Explore

4-<WBR>(Dimethylamino)<WBR>pyridine

ReagentPlus®, ≥99%

<I>p</I>-Toluenesulfonic acid monohydrate

ACS reagent, ≥98.5%

Pyridine
270970Pyridine

anhydrous, 99.8%

Triethylamine

≥99%

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
1
RTECS 
DB8929000
Flash Point(F) 
262.4 °F
Flash Point(C) 
128 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Immobilization of collagen onto polymer surfaces having hydroxyl groups. Tabata Y, Lonikar SV, Horii F, et al. Biomaterials 7(3), 234-8, (1986)

Loading...


Single hollow fiber SLM extraction of polyamines followed by tosyl chloride derivatization and HPLC determination. Dziarkowska K, Jönsson JA, and Wieczorek PP Anal. Chim. Acta 606(2), 184-93, (2008)

Loading...


Para-toluenesulfonyl chloride. Am. Ind. Hyg. Assoc. J. 43(10), B53-4, (1982)

Loading...


[Surface charge of mouse neuroblastoma cells and changes during growth and morphologic differentiation of the cell population]. Dolgaia EV, Mironov SL, and Pogorelaia NKh Neirofiziologiia 17(2), 168-74, (1985)

Loading...


Contribution of different scintigraphic techniques to the management of medullary thyroid carcinoma. Sandrock D, Blossey HC, Steinroeder M, et al. Henry Ford Hosp. Med. J. 37(3-4), 173-4, (1989)

Loading...


Copper(II)-complex directed regioselective mono-p-toluenesulfonylation of cyclomaltoheptaose at a primary hydroxyl group position: an NMR and molecular dynamics-aided design. Law H, Benito JM, Fernández JM, et al. J. Phys. Chem. B 115(23), 7524-32, (2011)

Loading...


Determination of glyphosate and its metabolite aminomethylphosphonic acid in fruit juices using supported-liquid membrane preconcentration method with high-performance liquid chromatography and UV detection after derivatization with p-toluenesulphonyl chloride. Khrolenko MV and Wieczorek PP J. Chromatogr. A 1093(1-2), 111-7, (2005)

Loading...


Isothiocyanates from tosyl chloride mediated decomposition of in situ generated dithiocarbamic acid salts. Wong R and Dolman SJ J. Org. Chem. 72(10), 3969-71, (2007)

Loading...


Kinetics and mechanism of imidazole-catalyzed acylation of cellulose in LiCl/N,N-dimethylacetamide. Nawaz H, Pires PA, and El Seoud OA Carbohydr. Polym. 92(2), 997-1005, (2013)

Loading...


Application of the dissociative electron transfer theory and its extension to the case of in-cage interactions in the electrochemical reduction of arene sulfonyl chlorides. Houmam A and Hamed EM Phys. Chem. Chem. Phys. 14(1), 113-24, (2012)

Loading...


High-performance liquid chromatographic determination of glyphosate and (aminomethyl)phosphonic acid in human serum after conversion into p-toluenesulphonyl derivatives. Tomita M, Okuyama T, Watanabe S, et al. J. Chromatogr. A 566(1), 239-43, (1991)

Loading...


Surface modifications of alginate/poly(L-lysine) microcapsular membranes with poly(ethylene glycol) and poly(vinyl alcohol). Kung IM, Wang FF, Chang YC, et al. Biomaterials 16(8), 649-55, (1995)

Loading...


Synthesis and properties of optically active nanostructured polymers bearing amino acid moieties by direct polycondensation of 4,4'-thiobis(2-tert-butyl-5-methylphenol) with chiral diacids. Mallakpour S and Soltanian S Amino Acids 42(6), 2187-94, (2012)

Loading...


[Degradation reactions of oral antidiabetics. 2. Reaction of arylsulfonylureas with 4-toluenesulfonyl chloride]. Egg H, Leibetseder H, Patzak A, et al. Arch. Pharm. (Weinheim) 320(3), 253-7, (1987)

Loading...


Studies of the synthesis of biomarkers. VII. Synthesis of 5 alpha-(17R,20R)-14,15-secocholestane. Li TS, Li YL, and Liang XT Steroids 55(6), 263-5, (1990)

Loading...


Occupational contact dermatitis from tosyl chloride in a chemist. Watsky KL, Reynolds K, Berube D, et al. Contact Dermatitis 29(4), 211-2, (1993)

Loading...


Direct regioselective 2-O-(p-toluenesulfonylation) of sucrose. Teranishi K Carbohydr. Res. 337(7), 613-9, (2002)

Loading...


Treatment of alcohols with tosyl chloride does not always lead to the formation of tosylates. Ding R, He Y, Wang X, et al. Molecules 16(7), 5665-73, (2011)

Loading...


Development and validation of an HPLC-DAD method for quantification of bornesitol in extracts from Hancornia speciosa leaves after derivatization with p-toluenesulfonyl chloride. Pereira AB, Veríssimo TM, Oliveira MA, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 887-888, 133-7, (2012)

Loading...


DNase activity of micrococcal endonuclease insolubilized on corn cob. Sánchez-Montero JM, Sinisterra JV, and Ballesteros A Appl. Biochem. Biotechnol. 22(2), 205-14, (1989)

Loading...


The analysis of platelet reaction, utilizing various inhibitors of platelet aggregation. Kambayashi J, Nomura T, Fujimoto T, et al. Nippon. Ketsueki Gakkai Zasshi 43(6), 1145-54, (1980)

Loading...


Synthesis of 1,2-unsaturated pyranosylphosphonate nucleosides from 3,4,6-tri-O-acetyl-D-glycal. Ismail AH Pharmazie 56(7), 534-5, (2001)

Loading...


(4S,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one. Robinson PD, Hua DH, Good LA, et al. Acta Crystallogr. C 49 ( Pt 6), 1238-40, (1993)

Loading...


Chemical modification of milk xanthine oxidase with different modifiers. Aboul-Enein HY, Refaie MO, El-Gazzar H, et al. Prep Biochem Biotechnol. 33(3), 173-87, (2003)

Loading...


Tosyl chloride. Schmidt RJ Contact Dermatitis 32(2), 124-5, (1995)

Loading...


Merck 14,9534

Beil. 11,IV,375

Fieser 1,1179 / Fieser 3,292 / Fieser 4,510 / Fieser 5,676 / Fieser 6,598 / Fieser 8,489 / Fieser 9,472 / Fieser 11,536 / Fieser 13,313

Aldrich MSDS 1, 1715:C / Corp MSDS 1 (2), 3367:C / FT-IR 1 (2), 520:B / FT-IR 2 (2), 3276:C / FT-NMR 1 (2), 1620:B / IR-Spectra (2), 1018:B / IR-Spectra (3), 1169:D / NMR-Reference 2 (2), 839:C / RegBook 1 (2), 2229:I / Sax 6, 2591 / Sigma FT-IR 1 (2), 932:D / Structure Index 1, 350:B:1

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?