89797 Fluka

Ursolic acid

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 3β-Hydroxy-12-ursen-28-ic acid

  • CAS Number 77-52-1

  • Empirical Formula (Hill Notation) C30H48O3

  • Molecular Weight 456.70

  •  Beilstein Registry Number 2228563

  •  EC Number 201-034-0

  •  MDL number MFCD00009621

  •  PubChem Substance ID 24889274

  • Popular Documents:  FTNMR (PDF)  

Price and Availability

Customers Also Viewed

Betulinic acid

≥98% (HPLC)

Maslinic acid

analytical standard

Oleanolic acid

analytical standard

Oleanolic acid

≥97%

Ursolic acid

≥90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?