Synonym: Certified Multielement Ion Chromatography Anion Standard Solution
EC Number 231-791-2
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs), |
| grade | for ion chromatography |
| shelf life | (limited shelf life, expiry date on the label) |
Certified primary reference material for Ion Chromatography; high purity starting materials certified by EMPA/BAM
Produced with certified high purity materials (NaF, NaCl, NaBr, NaNO3, Na2HPO4, Na2SO4) and ultrapure water (18 MOhm, 0.22 μm filtered)
Customers Also Viewed
TraceCERT®, in 0.1% nitric acid
analytical standard
for ion chromatography, Reference material traceable to SI, 10.0 mg/kg ± 0.2% Li, Na, K, Mg, Ca each cation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?