90161 Fluka

(2,2′)-Furildioxime

for spectrophotometric det. of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII), U(VI), ≥97.0% (sum of isomers, HPLC)

DOWNLOAD MSDS (PDF)

Synonym: Bis(2-furyl)glyoxime

  • CAS Number 522-27-0

  • Empirical Formula (Hill Notation) C10H8N2O4

  • Molecular Weight 220.18

  •  EC Number 208-326-7

  •  MDL number MFCD00003247

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
description   mixture of isomers
assay   ≥97.0% (sum of isomers, HPLC)

Description

Application

Reagent for the spectrophotometric determination of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII) and U(VI), a review1

Price and Availability

Customers Also Viewed

Furil
138029Furil

98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?