EMAIL THIS PAGE TO A FRIEND

90370 Fluka

Triethyl borate

technical, ≥98.0% (GC)

Synonym: Boric acid triethyl ester, Boron ethoxide, Triethoxyborane

Purchase

Properties

vapor density   5.04 (vs air)
grade   technical
assay   ≥98.0% (GC)
form   liquid
refractive index   n20/D 1.374(lit.)
  n20/D 1.376
bp   117-118 °C(lit.)
density   0.858 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Tributyl borate

≥99.0% (T)

Triethylborane

≥95%

Trimethyl borate

99.999% (trace metal basis)

Trimethyl borate

≥98%

Safety & Documentation

Safety Information

RIDADR 
UN 1176 3/PG 2
WGK Germany 
2
RTECS 
ED5075000
Flash Point(F) 
52 °F
Flash Point(C) 
11 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 1,IV,1365

Aldrich MSDS 1, 1750:C / Corp MSDS 1 (2), 3430:C / FT-NMR 1 (3), 725:A / IR-Spectra (3), 1550:D / IR-Spectra (2), 1325:E / NMR-Reference 2 (2), 1010:B / RegBook 1 (2), 3023:D / Sax 6, 2639 / Structure Index 1, 478:C:3 / Vapor Phase 3, 1676:C

Related Products

replaced by

Product #

Description

Add to Cart

T59307 Triethyl borate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?