EMAIL THIS PAGE TO A FRIEND

90390 Fluka

Triethylene glycol

puriss., anhydrous, ≥99.0% (GC)

Synonym: Triglycol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
vapor density   5.2 (vs air)
vapor pressure   <0.01 mmHg ( 20 °C)
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   699 °F
expl. lim.   9.2 %
impurities   ≤0.08% water
refractive index   n20/D 1.455(lit.)
  n20/D 1.456
bp   125-127 °C/0.1 mmHg(lit.)
mp   −7 °C(lit.)
density   1.124 g/mL at 20 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Triethylene glycol

BioUltra, anhydrous, ≥99.0% (GC)

Triethylene glycol

ReagentPlus®, 99%

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
YE4550000
Flash Point(F) 
330.8 °F
Flash Point(C) 
166 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

pH-dependent, thermosensitive polymeric nanocarriers for drug delivery to solid tumors. Chen CY, Kim TH, Wu WC, et al. Biomaterials 34(18), 4501-9, (2013)

Loading...


Photocontrolled nanoparticles for on-demand release of proteins. Azagarsamy MA, Alge DL, Radhakrishnan SJ, et al. Biomacromolecules 13(8), 2219-24, (2012)

Loading...


Tuning stamp surface energy for soft lithography of polar molecules to fabricate bioactive small-molecule microarrays. Vaish A, Shuster MJ, Cheunkar S, et al. Small 7(10), 1471-9, (2011)

Loading...


Supramolecular construction of vesicles based on core-substituted naphthalene diimide appended with triethyleneglycol motifs. Bhosale SV, Jani CH, Lalander CH, et al. Chem. Commun. (Camb.) 47(29), 8226-8, (2011)

Loading...


Bifunctional dendronized cellulose surfaces as biosensors. Montanez MI, Hed Y, Utsel S, et al. Biomacromolecules 12(6), 2114-25, (2011)

Loading...


Polymerization of human hemoglobin using the crosslinker 1,11-bis(maleimido)triethylene glycol for use as an oxygen carrier. Zhang N and Palmer AF Biotechnol. Prog. 26(5), 1481-5, (2010)

Loading...


The impact of three strains of oral bacteria on the surface and mechanical properties of a dental resin material. Gregson KS, Shih H, and Gregory RL Clin. Oral Investig. 16(4), 1095-103, (2012)

Loading...


A novel water-soluble NHC-Pd polymer: an efficient and recyclable catalyst for the Suzuki coupling of aryl chlorides in water at room temperature. Karimi B and Fadavi Akhavan P Chem. Commun. (Camb.) 47(27), 7686-8, (2011)

Loading...


Diethylene glycol in health products sold over-the-counter and imported from Asian countries. Schier JG, Barr DB, Li Z, et al. J. Med. Toxicol. 7(1), 33-8, (2011)

Loading...


Dendrimers as potential inhibitors of the dimerization of the capsid protein of HIV-1. Doménech R, Abian O, Bocanegra R, et al. Biomacromolecules 11(8), 2069-78, (2010)

Loading...


Oxidation of tertiary amine-derivatized surfaces to control protein adhesion. Dobrzanska DA, Cooper AL, Dowson CG, et al. Langmuir 29(9), 2961-70, (2013)

Loading...


Separation of preferential interaction and excluded volume effects on DNA duplex and hairpin stability. Knowles DB, LaCroix AS, Deines NF, et al. Proc. Natl. Acad. Sci. U. S. A. 108(31), 12699-704, (2011)

Loading...


Aggregation behavior modulation of 1-dodecyl-3-methylimidazolium bromide by organic solvents in aqueous solution. Wang J, Zhang L, Wang H, et al. J. Phys. Chem. B 115(17), 4955-62, (2011)

Loading...


Dendrimers reduce toxicity of Aβ 1-28 peptide during aggregation and accelerate fibril formation. Klajnert B, Wasiak T, Ionov M, et al. Nanomedicine 8(8), 1372-8, (2012)

Loading...


Effects of a PEG additive on the biomolecular interactions of self-assembled dendron nanostructures. Barnard A, Calderon M, Tschiche A, et al. Org. Biomol. Chem. 10(42), 8403-9, (2012)

Loading...


Comprehensive comparison of two new biodegradable gene carriers. Zhao D, Gong T, Zhu D, et al. Int. J. Pharm. 413(1-2), 260-70, (2011)

Loading...


One-step detection of galectins on hybrid monolayer surface with protruding lactoside. Yoshioka K, Sato Y, Murakami T, et al. Anal. Chem. 82(4), 1175-8, (2010)

Loading...


Use of a polyol liquid collection medium to obtain ultrasmall magnetic nanoparticles by laser pyrolysis. Martínez G, Malumbres A, Mallada R, et al. Nanotechnology 23(42), 425605, (2012)

Loading...


Study and fabrication of europium picrate triethylene glycol complex. Saleh MI, Kusrini E, Mohd Sarjidan MA, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 78(1), 52-8, (2011)

Loading...


Construction of a long range p/n heterojunction with a pair of nanometre-wide continuous D/A phases. Dong L, Li W, and Li WS Nanoscale 3(9), 3447-61, (2011)

Loading...


Efficient multigram synthesis of the repeating unit of gallic acid-triethylene glycol dendrimers. Amaral SP, Fernandez-Villamarin M, Correa J, et al. Org. Lett. 13(17), 4522-5, (2011)

Loading...


Oriented immobilization of prion protein demonstrated via precise interfacial nanostructure measurements. Sanavio B, Scaini D, Grunwald C, et al. ACS Nano 4(11), 6607-16, (2010)

Loading...


Investigation of crystal structure influence on spectroscopic and photoluminescent properties of terbium-picrate triethylene glycol complex. Kusrini E, Saleh MI, Adnan R, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 98, 322-8, (2012)

Loading...


Stabilizing labile DNA-protein complexes in polyacrylamide gels. Sidorova NY, Hung S, and Rau DC Electrophoresis 31(4), 648-53, (2010)

Loading...


Phytoremediation of ethylene glycol and its derivatives by the burhead plant (Echinodorus cordifolius (L.)): effect of molecular size. Teamkao P and Thiravetyan P Chemosphere 81(9), 1069-74, (2010)

Loading...


Bifunctional tri(ethylene glycol) alkanethiol monolayer modified gold electrode for on-chip electrochemical immunoassay of pg level leptin. Nishimura T, Sato Y, Tanaka M, et al. Anal. Sci. 27(5), 465, (2011)

Loading...


Triethylene glycol HO(CH2CH2O)3H. Ballantyne B and Snellings WM J. Appl. Toxicol. 27(3), 291-9, (2007)

Loading...


Final report on the safety assessment of Triethylene Glycol and PEG-4. Int. J. Toxicol. 25 Suppl 2, 121-38, (2006)

Loading...


Merck 14,9670

Beil. 1,IV,2400

Aldrich MSDS 1, 1750:D / Corp MSDS 1 (2), 3430:D / FT-IR 1 (1), 226:B / FT-IR 2 (1), 331:A / FT-NMR 1 (1), 345:A / IR-Spectra (2), 123:C / IR-Spectra (3), 134:G / NMR-Reference 2 (1), 192:C / RegBook 1 (1), 227:D / Sax 6, 2639 / Structure Index 1, 31:B:6 / Vapor Phase 3, 297:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?