Synonym:
1,3,4,6,7,8-
MDL number MFCD00145662
PubChem Substance ID 24889384
| Related Categories | Bases, Chemical Synthesis, Organic Bases, Solid Supported Synthesis, Specialty Synthesis, |
| extent of labeling | ~2.6 mmol/g base loading |
| matrix | crosslinked with 2% DVB |
| particle size | 200-400 mesh |
Polymer-supported, strong, hindered pentasubstituted guanidine base
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?