90605 Fluka

1,5,7-Triazabicyclo[4.4.0]dec-5-ene

purum, ≥98.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, TBD

  • CAS Number 5807-14-7

  • Empirical Formula (Hill Notation) C7H13N3

  • Molecular Weight 139.20

  •  Beilstein Registry Number 3242

  •  EC Number 227-367-1

  •  MDL number MFCD00043003

  •  PubChem Substance ID 24889385

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, N-Containing, Others More...
grade   purum
assay   ≥98.0% (T)
mp   125-130 °C(lit.)
  125-132 °C

Description

Other Notes

Guanidine base which is about 100 times more basic than tetramethylguanidine1; Powerful base for tautomerizing pyrrocorphins and for other reactions2,3

Price and Availability

Customers Also Viewed

1,3,4,6,7,8-<WBR>Hexahydro-<WBR>2<I>H</I>-pyrimido[1,2-<I>a</I>]pyrimidine, polymer-bound

200-400 mesh, extent of labeling: 2.6 mmol/g loading, 2 % cross-linked with divinylbenzene

1,5,7-<WBR>Triazabicyclo[4.4.0]<WBR>dec-<WBR>5-<WBR>ene bound to polystyrene

extent of labeling: ~2.6 mmol/g base loading

1,8-<WBR>Diazabicyclo[5.4.0]<WBR>undec-<WBR>7-<WBR>ene

puriss., ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?