Synonym:
Melamine-
MDL number MFCD00197912
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, M, |
| form | aqueous suspension (unless otherwise indicated) |
| concentration | 10% (solids) |
| particle size | 3 μm std dev <0.1 μm, coeff var <3% |
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.
Density: 1.51 g/cm3
Heat resistance <300 °C
Chemical resistance: acids, bases, organic solvents.
Monodispers melamin resin particles are characterized by high uniformity and narrow size distribution. They carry functional methylol groups, which can be used for covalent coupling of biological active compounds.
Additional advantages of melamin resin particles are:
• hydrophilic surfaces
• high degree of cross linking and high pressure resistance
• long term stability in aqueous media (freezing and thawing does not damage them)
• lyophilization and resuspension in aqueous media does not damage them (powders are available on request)
• no swelling in organic solvents
Customers Also Viewed
size: 3 μm
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?