EMAIL THIS PAGE TO A FRIEND

91446 Fluka

Pentadecanoic acid

analytical standard

Synonym: C15:0, NSC 28486, Pentadecylic acid

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Fatty Acids, Free Fatty Acids,
grade   analytical standard
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
bp   257 °C/100 mmHg(lit.)
mp   51-53 °C(lit.)
  51-55 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Fatty Acid and Sterol Extraction Kits

Customers Also Viewed

Methyl pentadecanoate

analytical standard

Pentadecanoic acid

~99% (capillary GC)

Pentadecanoic acid

≥99%

Tridecanoic acid

analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Fatty acid composition of adipose tissue and serum lipids are valid biological markers of dairy fat intake in men. Wolk A, Furuheim M, and Vessby B J. Nutr. 131(3), 828-33, (2001)

Loading...


HMDB: a knowledgebase for the human metabolome. Wishart DS, Knox C, Guo AC, et al. Nucleic Acids Res. 37(Database issue), D603-10, (2009)

Loading...


Evaluation of a biological marker of dairy fat intake. Wolk A, Vessby B, Ljung H, et al. Am. J. Clin. Nutr. 68(2), 291-5, (1998)

Loading...


The human serum metabolome. Psychogios N, Hau DD, Peng J, et al. PLoS ONE 6(2), e16957, (2011)

Loading...


Protection from obesity and insulin resistance in mice overexpressing human apolipoprotein C1. Jong MC, Voshol PJ, Muurling M, et al. Diabetes 50(12), 2779-85, (2001)

Loading...


Lipidomics reveals a remarkable diversity of lipids in human plasma. Quehenberger O, Armando AM, Brown AH, et al. J. Lipid Res. 51(11), 3299-305, (2010)

Loading...


Incremental predictive value of myocardial scintigraphy with 123I-BMIPP in patients with acute myocardial infarction treated with primary percutaneous coronary intervention. Nanasato M, Hirayama H, Ando A, et al. Eur. J. Nucl. Med. Mol. Imaging 31(11), 1512-21, (2004)

Loading...


Interference by naturally occurring fatty acids in a noncellular enzyme-based aromatase bioassay. Marcy J Balunas et al J. Nat. Prod. 69, 700-3, (2006)

Loading...


Prediction of functional improvement of ischemic myocardium with (123I-BMIPP SPECT and 99mTc-tetrofosmin SPECT imaging: a study of patients with large acute myocardial infarction and receiving revascularization therapy. Seki H, Toyama T, Higuchi K, et al. Circ. J. 69(3), 311-9, (2005)

Loading...


Contractile reserve, thallium-201 reverse redistribution and mismatch between perfusion and metabolism in reperfused infarct-related myocardium with delayed and incomplete functional recovery. Akutsu Y, Kodama Y, Nishimura H, et al. Jpn. Heart J. 45(5), 739-48, (2004)

Loading...


Detecting viable myocardium and predicting functional improvement: comparisons of positron emission tomography, rest-redistribution thallium-201 single-photon emission computed tomography (SPECT), exercise thallium-201 reinjection SPECT, I-123 BMIPP SPECT and dobutamine stress echocardiography. Tani T, Teragaki M, Watanabe H, et al. Circ. J. 68(10), 950-7, (2004)

Loading...


[Diagnosis and therapy of postoperative chyloperitoneum]. Bauwens K, Jacobi CA, Gellert K, et al. Chirurg. 67(6), 658-60, (1996)

Loading...


Scintigraphic evaluation of cardiac metabolism in multicentric Castleman's disease. Matsuo S, Nakamura Y, Nakae I, et al. Intern. Med. 43(6), 490-2, (2004)

Loading...


Beil. 2,IV,1147

Aldrich MSDS 1, 1424:D / Corp MSDS 1 (2), 2698:B / FT-IR 2 (1), 759:D / FT-IR 1 (1), 484:D / FT-NMR 1 (1), 756:A / IR-Spectra (3), 285:H / IR-Spectra (2), 257:H / NMR-Reference 2 (1), 422:C / RegBook 1 (1), 535:I / Sax 6, 2128 / Sigma FT-IR 1 (1), 1454:A / Structure Index 1, 75:C:6 / Vapor Phase 3, 577:A

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?