Synonym: 3β-Hydroxy-20(29)-lupaene-28-oic acid
CAS Number 472-15-1
Empirical Formula (Hill Notation) C30H48O3
Molecular Weight 456.70
EC Number 207-448-8
MDL number MFCD00009619
| Related Categories | A - D, Alphabetical, Analytical Standards, Analytical/Chromatography, Betula, |
| grade | analytical standard |
| assay | ≥97.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 295-298 °C (dec.)(lit.) |
| storage temp. | 2-8°C |
Betulinic acid, a pentacyclic triterpene, selectively induces apoptosis in tumor cells by directly activating the mitochondrial pathway of apoptosis through a p53- and CD95-independent mechanism.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?