Synonym:
1-
CAS Number 6138-41-6
Empirical Formula (Hill Notation) C7H7NO2 · HCl
Molecular Weight 173.60
Beilstein Registry Number 3734169
EC Number 228-119-5
MDL number MFCD00077250
| Related Categories | Building Blocks, C7, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More... |
| grade | purum |
| assay | ≥98.0% (AT) |
| mp | ~260 °C (dec.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?