91928 Fluka

2′,4′,6′-Trihydroxyacetophenone monohydrate

matrix substance for MALDI-MS, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: 2-Acetylphloroglucinol, THAP

  • CAS Number 480-66-0

  • Linear Formula (HO)3C6H2COCH3·H2O

  • Molecular Weight 186.16

  •  Beilstein Registry Number 1911197

  •  EC Number 207-556-5

  •  MDL number MFCD00149091

  •  PubChem Substance ID 24889584

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Glycan Labeling and Analysis, Glycobiology, Mass Spectrometry (MS),
grade   matrix substance for MALDI-MS
assay   ≥99.5% (HPLC)
  ≥99.5%
color   light yellow
mp   218-222 °C
  219-221 °C(lit.)
cation traces   Ca: ≤200 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
suitability   in accordance for UV test
analyte suitability   glycans
  oligonucleotides
  peptides
  proteins

Description

Application

Used in MALDI analysis of acidic glycans and glyco­peptides in negative ion mode. Lower limit of detection than 6-aza-2-thiothimidine.1

Protocols & Applications

Methods of glycosylated protein analysis by Mass Spectrometry

Price and Availability

Customers Also Viewed

2,5-Dihydroxybenzoic acid

matrix substance for MALDI-MS, >99.0% (HPLC)

3-Hydroxypicolinic acid

matrix substance for MALDI-MS, ≥99.0% (HPLC)

6-Aza-2-thiothymine

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?