92360 Fluka

Chlorotrimethylsilane

puriss., ≥99.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: TMCS, Trimethylchlorosilane, Trimethylsilyl chloride

  • CAS Number 75-77-4

  • Linear Formula (CH3)3SiCl

  • Molecular Weight 108.64

  •  Beilstein Registry Number 1209232

  •  EC Number 200-900-5

  •  MDL number MFCD00000502

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents GC,
vapor density   3.7 (vs air)
vapor pressure   100 mmHg ( 25 °C)
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   752 °F
expl. lim.   6.4 %
refractive index   n20/D 1.387(lit.)
bp   57 °C(lit.)
mp   −40 °C(lit.)
density   0.856 g/mL at 25 °C(lit.)

Description

Other Notes

Silylating agent1,2

Price and Availability

Customers Also Viewed

Chlorotrimethylsilane

purified by redistillation, ≥99%

Chlorotrimethylsilane

produced by Wacker, ≥99.0% (GC)

Chlorotrimethylsilane

≥98.0% (GC)

Hexamethyldisilazane

for GC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?