Synonym: Lup-20(29)-ene-3β,28-diol
CAS Number 473-98-3
Empirical Formula (Hill Notation) C30H50O2
Molecular Weight 442.72
EC Number 207-475-5
MDL number MFCD00016802
| Related Categories | A - D, Alphabetical, Analytical Standards, Analytical/Chromatography, Betula, |
| grade | analytical standard |
| assay | ≥97.5% (TLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 256-257 °C(lit.) |
Bottomless glass bottle. Contents are inside inserted fused cone.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?