• USA Home
  • 93510 - (S)-Trolox methyl ether

EMAIL THIS PAGE TO A FRIEND

93510 Fluka

(S)-Trolox methyl ether

for chiral derivatization, ≥98.0%

Synonym: (S)-6-Methoxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

  • CAS Number 135806-59-6

  • Empirical Formula (Hill Notation) C15H20O4

  • Molecular Weight 264.32

  •  Beilstein Registry Number 5481541

  •  MDL number MFCD00171453

  •  PubChem Substance ID 24889901

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chiral, Chiral Derivatization Reagents, Chiral LC Derivatization Reagents,
grade   for chiral derivatization
assay   ≥98.0% (sum of enantiomers, HPLC)
  ≥98.0%
optical activity   [α]20/D −69±2°, c = 1% in ethanol
optical purity   enantiomeric ratio: ≥99.5:0.5 (HPLC)
mp   143-145 °C

Description

Other Notes

Chiral derivatizing agent for alcohols; some methyl substituted primary alcohols which are difficult to separate otherwise, can also be separated1,2

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. W. Walther et al. Chimia 45, 121, (1991)

2. W. Walther et al. J. Microcol. Sep. 4, 45, (1992)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?