93595 Fluka

Trypan Blue solution

0.4%, for microscopy

DOWNLOAD MSDS (PDF)
  • CAS Number 72-57-1

  • Empirical Formula (Hill Notation) C34H24N6Na4O14S4

  • Molecular Weight 960.81

  •  Beilstein Registry Number 4360496

  •  MDL number MFCD00003969

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Microscopy Reagents, Stains and Dyes
grade   for microscopy
shelf life   (limited shelf life, expiry date on the label)
concentration   0.4%
density   1.007 g/mL at 20 °C

Description

Physical form

0.4% in 0.81% sodium chloride; 0.06% potassium phosphate, dibasic

Price and Availability

Customers Also Viewed

Dimethyl sulfoxide

Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%

Trypan Blue

Dye content 60 %

Trypan Blue

powder, BioReagent, suitable for cell culture

Trypan Blue solution

0.4%, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?