93894 Fluka

(−)-Epigallocatechin gallate

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, EGCG

  • CAS Number 989-51-5

  • Empirical Formula (Hill Notation) C22H18O11

  • Molecular Weight 458.37

  •  MDL number MFCD00075940

Properties

Related Categories Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, E - H,
grade   analytical standard
assay   ≥98.0% (HPLC)
shelf life   (limited shelf life, expiry date on the label)
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

(−)-Epicatechin gallate

≥98% (HPLC), from green tea

(−)-Epigallocatechin

analytical standard

(−)-Epigallocatechin

≥95% (HPLC), from green tea

(−)-Epigallocatechin gallate

purum, ≥97.0% (HPLC)

(−)-Epigallocatechin gallate

≥95%, from green tea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?