• USA Home
  • 94270 - Uranyl nitrate hexahydrate

EMAIL THIS PAGE TO A FRIEND
94270 Fluka

Uranyl nitrate hexahydrate

puriss. p.a., ACS reagent, 98.0-102.0%

Purchase

Properties

grade   ACS reagent
  puriss. p.a.
assay   98.0-102% (RT)
  98.0-102.0%
extent of labeling   1.23E4 Bq/g specific activity
anion traces   chloride (Cl-): ≤30 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Fe: ≤10 mg/kg

Description

Other Notes

Sales restrictions may apply

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1477 5.1/PG 2
WGK Germany 
3
RTECS 
YR3850000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?