95038 Fluka

Propyl 4-hydroxybenzoate sodium salt

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: Propylis parahydroxybenzoas natricum, Sodium propyl 4-hydroxybenzoate

  • CAS Number 35285-69-9

  • Empirical Formula (Hill Notation) C10H11NaO3

  • Molecular Weight 202.18

  •  Beilstein Registry Number 5183350

  •  EC Number 252-488-1

  •  MDL number MFCD00016477

  •  PubChem Substance ID 24890084

Properties

Related Categories Pharmacopoeia, Pharmacopoeia A-Z More...
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?