Synonym: Peptone from casein, tryptic digest, Peptone from casein
CAS Number 91079-40-2
| Related Categories | Base Ingredients for Media, Microbiology, Protein Sources (Peptones & Extracts), Protein Sources (milk derived), Protein Sources A to Z More... |
| grade | for biotechnological purposes |
| impurities | ≥12% total nitrogen (N) |
| ≥3.5% amino N | |
| ign. residue | ≤17.0% |
| loss | ≤6.0% loss on drying |
| pH | 6.0-7.5 (25 °C, 2% in H2O) |
| solubility | H2O: soluble2%, clear, faintly to slightly yellow |
A rich source of amino acids and peptides recommended for culture media in analytical microbiology and in industrial fermentation.
Customers Also Viewed
for biotechnological purposes
for microbiology
Microbiologically tested.
Pancreatic digest of casein
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?