• USA Home
  • 95137 - 1-Butyl-3-methylimidazolium bromide

EMAIL THIS PAGE TO A FRIEND

95137 Fluka

1-Butyl-3-methylimidazolium bromide

>97.0% (HPLC)

Synonym: BMIMBr

Purchase

Properties

Related Categories Chemical Synthesis, Imidazolium, Ionic Liquids, Specialty Synthesis
assay   >97.0% (HPLC)
impurities   ≤1% water

Description

Other Notes

Ionic liquid used in a Heck reaction1

Packaging

5, 50 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. L. Xu et al., et al. Organometallics 19, 1123, (2000)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?