Synonym: Acid blue 93, Aniline blue water soluble, Cotton blue, Poirriers blue, Water blue
CAS Number 28983-56-4
Empirical Formula (Hill Notation) C37H27N3Na2O9S3
Molecular Weight 799.80
Colour Index Number 42780
EC Number 249-352-9
MDL number MFCD00058509
| Related Categories | Analytical Reagents, Analytical/Chromatography, Microscopy Reagents, Stains and Dyes |
| grade | for microscopy (Bot., Hist.) |
| indicator (pH 9.4-14.0) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?