95670 Fluka

m-Xylene

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,3-Dimethylbenzene

  • CAS Number 108-38-3

  • Linear Formula C6H4(CH3)2

  • Molecular Weight 106.17

  •  Beilstein Registry Number 605441

  •  EC Number 203-576-3

  •  MDL number MFCD00008536

  •  PubChem Substance ID 24890183

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
vapor density   3.7 (vs air)
assay   ≥99.5% (GC)
autoignition temp.   982 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   7 %
refractive index   n20/D 1.497(lit.)
  n20/D 1.497
bp   138-139 °C(lit.)
mp   −48 °C(lit.)
density   0.868 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

<I>m</I>-Xylene

puriss. p.a., ≥99.0% (GC)

<I>m</I>-Xylene
296325m-Xylene

anhydrous, ≥99%

<I>m</I>-Xylene
185566m-Xylene

ReagentPlus®, 99%

<I>o</I>-Xylene

analytical standard

<I>p</I>-Xylene

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?